Assay Details
Target Gene Details
Entrez Gene ID: | 11998 |
Gene Name: | arginine vasopressin |
Gene Aliases: |
Vp, Vsp |
Location: |
Chr.2:130580677-130582541 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Avp | NM_009732.1 | NP_033862.1 | ||
| AK138179.1 | 1 | 57 | BAE23570.1 | |
| BC051997.1 | AAH51997.1 |
More Information
Set Membership: |
|

Genomic Map