Assay Details
Target Gene Details
Entrez Gene ID: | 271887 |
Gene Name: | RBBP8 N-terminal like |
Gene Aliases: |
4732416F18 |
Location: |
Chr.2:180275246-180289900 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbbp8nl | NM_173031.3 | NP_766619.2 | ||
| AK028610.1 | BAC26032.1 | |||
| BY714078.1 |
More Information
Set Membership: |
|

Genomic Map