Assay Details
Target Gene Details
Entrez Gene ID: | 69379 |
Gene Name: | complement component 8, gamma polypeptide |
Gene Aliases: |
- |
Location: |
Chr.2:25498650-25501926 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8g | NM_001271777.1 | NP_001258706.1 | ||
| NM_027062.2 | NP_081338.1 | |||
| AK005962.1 | BAB24341.1 | |||
| BC019967.1 | AAH19967.1 | |||
| BY090016.1 |

Genomic Map