Assay Details
Target Gene Details
Entrez Gene ID: | 56336 |
Gene Name: | UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 5 |
Gene Aliases: |
9430078I07Rik, AW049941, AW539721 |
Location: |
Chr.2:167298444-167349178 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4galt5 | NM_019835.2 | 2 | 332 | NP_062809.2 |
| AB004786.1 | 2 | 333 | BAA94791.1 | |
| AF142673.1 | 2 | 186 | AAF22223.1 | |
| BE981379.1 | 2 | 202 | ||
| CJ134591.1 | 2 | 333 |
More Information
Set Membership: |
|

Genomic Map