Assay Details
Target Gene Details
Entrez Gene ID: | 329547 |
Gene Name: | bactericidal permeablility increasing protein |
Gene Aliases: |
9230105K17Rik, Bpifd1 |
Location: |
Chr.2:158258091-158284533 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bpi | NM_177850.3 | 10 | 1095 | NP_808518.1 |
| AK033770.1 | 10 | 1097 | BAC28468.1 | |
| AY363993.1 | 10 | 1093 | AAR13289.1 | |
| AY648037.1 | 10 | 1095 | AAV65841.1 | |
| AY853179.1 | 10 | 1080 | AAW50819.1 | |
| BC118049.1 | 10 | 1089 | ||
| BC118504.1 | 10 | 1089 |

Genomic Map