Assay Details
Target Gene Details
Entrez Gene ID: | 227723 |
Gene Name: | proline-rich coiled-coil 2B |
Gene Aliases: |
5830434P21Rik, AI173903, Bat2l, Bat2l1, D430039P21, mKIAA0515 |
Location: |
Chr.2:32151148-32234537 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prrc2b | NM_001159634.1 | NP_001153106.1 | ||
| NM_172661.3 | NP_766249.2 | |||
| AK052511.1 | BAC35022.1 | |||
| AK147271.1 | ||||
| AK147677.1 | ||||
| BC055116.1 | AAH55116.1 |
More Information
Set Membership: |
|

Genomic Map