Assay Details
Target Gene Details
Entrez Gene ID: | 66557 |
Gene Name: | BPI fold containing family B, member 2 |
Gene Aliases: |
2310034L21Rik, 2310069A01Rik, AV089125, Bpil1, LPLUNC2, RYSR |
Location: |
Chr.2:153875045-153895270 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bpifb2 | NM_025631.3 | NP_079907.2 | ||
| AK009346.1 | ||||
| AK009504.1 | ||||
| AK009619.1 | BAB26395.1 | |||
| AK009754.1 | BAB26479.1 | |||
| AK009776.1 | ||||
| AK009794.1 | ||||
| AK009871.1 | ||||
| AK010130.1 | ||||
| AK010154.1 | ||||
| AK010157.1 | ||||
| AK010215.1 | ||||
| AK028156.1 | BAC25783.1 | |||
| AK028158.1 | BAC25784.1 | |||
| AK075874.1 | BAC36021.1 | |||
| AK160516.1 | BAE35837.1 | |||
| BC116681.1 | ||||
| BC116727.1 |

Genomic Map