Assay Details
Target Gene Details
Entrez Gene ID: | 332579 |
Gene Name: | caspase recruitment domain family, member 9 |
Gene Aliases: |
Gm782 |
Location: |
Chr.2:26352236-26360979 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Card9 | NM_001037747.1 | NP_001032836.1 | ||
| BC065797.1 | 1 | 174 | AAH65797.1 | |
| BC151023.1 |

Genomic Map