Assay Details
Target Gene Details
Entrez Gene ID: | 12298 |
Gene Name: | calcium channel, voltage-dependent, beta 4 subunit |
Gene Aliases: |
3110038O15Rik, Cchb4, lh |
Location: |
Chr.2:52428320-52676609 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacnb4 | NM_001037099.2 | NP_001032176.1 | ||
| NM_001285426.1 | NP_001272355.1 | |||
| NM_001285427.1 | NP_001272356.1 | |||
| NM_001285428.1 | NP_001272357.1 | |||
| NM_146123.3 | NP_666235.1 | |||
| AF039417.1 | AAB95484.1 | |||
| AK032311.1 | ||||
| AK038633.1 | BAC30073.1 | |||
| AK043850.1 | BAC31681.1 | |||
| AK079616.1 | BAC37703.1 | |||
| AK147338.1 | BAE27855.1 | |||
| AK158606.1 | BAE34578.1 | |||
| BC026479.1 | AAH26479.1 |
More Information
Set Membership: |
|

Genomic Map