Assay Details
Target Gene Details
Entrez Gene ID: | 12531 |
Gene Name: | cell division cycle 25B |
Gene Aliases: |
AI604853 |
Location: |
Chr.2:131186436-131198511 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc25b | NM_001111075.4 | 14 | 1686 | NP_001104545.1 |
| NM_023117.4 | 15 | 1764 | NP_075606.1 | |
| AK004836.1 | 14 | 1643 | BAB23603.1 | |
| AK043730.1 | 14 | 1677 | BAC31632.1 | |
| AK153903.1 | 15 | 1765 | BAE32245.1 | |
| AK157397.1 | 14 | 1644 | BAE34078.1 | |
| BC002287.1 | 6 | 551 | AAH02287.1 | |
| BC057568.1 | 14 | 1557 | AAH57568.1 | |
| S93521.1 | 15 | 1633 | AAB22026.1 |
More Information
Set Membership: |
|

Genomic Map