Assay Details
Target Gene Details
Entrez Gene ID: | 66205 |
Gene Name: | CD302 antigen |
Gene Aliases: |
1110055L24Rik, AI159627 |
Location: |
Chr.2:60251993-60284484 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd302 | NM_001290660.1 | 2 | 484 | NP_001277589.1 |
| NM_025422.4 | 2 | 484 | NP_079698.2 | |
| AI116405.1 | 2 | 484 | ||
| AK002812.1 | 2 | 125 | BAB22377.2 | |
| AK004267.1 | 2 | 420 | BAB23242.2 | |
| BC005501.2 | 1 | 46 | AAH05501.1 |

Genomic Map