Assay Details
Target Gene Details
Entrez Gene ID: | 110911 |
Gene Name: | CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 2 |
Gene Aliases: |
5730450N06Rik, 5730460C18Rik, AI854580, D2Wsu127e |
Location: |
Chr.2:132262609-132312050 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cds2 | NM_001291040.1 | 1 | 313 | NP_001277969.1 |
| NM_138651.7 | 1 | 313 | NP_619592.1 | |
| AK036328.1 | 1 | 164 | BAC29384.1 | |
| AK050589.1 | 1 | 221 | ||
| AK147541.1 | 1 | 205 | BAE27984.1 | |
| AK166001.1 | 1 | 164 | BAE38511.1 | |
| AK170888.1 | 1 | 202 | BAE42095.1 | |
| AY159802.1 | 1 | 236 | AAO17790.1 | |
| BC003852.1 | 1 | 219 | AAH03852.1 | |
| BC059776.1 | 1 | 116 | AAH59776.1 | |
| BC069879.1 | 1 | 192 | AAH69879.1 | |
| CJ112406.1 | 1 | 314 |
More Information
Set Membership: |
|

Genomic Map