Assay Details
Target Gene Details
Entrez Gene ID: | 110911 |
Gene Name: | CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 2 |
Gene Aliases: |
5730450N06Rik, 5730460C18Rik, AI854580, D2Wsu127e |
Location: |
Chr.2:132262609-132312050 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cds2 | NM_001291039.1 | NP_001277968.1 | ||
| NM_138651.7 | NP_619592.1 | |||
| AK036328.1 | BAC29384.1 | |||
| AK049257.1 | BAC33639.1 | |||
| AK147541.1 | BAE27984.1 | |||
| AK166001.1 | BAE38511.1 | |||
| AY159802.1 | AAO17790.1 | |||
| BC003852.1 | AAH03852.1 | |||
| BC069879.1 | AAH69879.1 |
More Information
Set Membership: |
|

Genomic Map