Assay Details
Target Gene Details
Entrez Gene ID: | 12521 |
Gene Name: | CD82 antigen |
Gene Aliases: |
AA682076, AL023070, C33, IA4, Kai1, Tspan27 |
Location: |
Chr.2:93419102-93462950 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd82 | NM_001136055.2 | NP_001129527.1 | ||
| NM_001271431.1 | NP_001258360.1 | |||
| AK135381.1 | BAE22511.1 | |||
| AK146912.1 | BAE27524.1 |
Target Gene Details
Entrez Gene ID: | 100038525 |
Gene Name: | predicted gene 10804 |
Gene Aliases: |
ENSMUSG00000075025 |
Location: |
Chr.2:93452819-93469874 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10804 | NR_040532.1 | |||
| AK143789.1 | BAE25541.1 |

Genomic Map