Assay Details
Target Gene Details
Entrez Gene ID: | 16328 |
Gene Name: | centrosomal protein 250 |
Gene Aliases: |
AW490617, B230210E21Rik, Cep2, Inmp |
Location: |
Chr.2:155956272-155998900 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 21 - Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep250 | NM_001129999.1 | NP_001123471.1 | ||
| NM_001130000.1 | NP_001123472.1 | |||
| NM_008383.3 | NP_032409.3 | |||
| NM_177217.3 | NP_796191.2 | |||
| AK145562.1 | ||||
| AK161443.1 | ||||
| BC150746.1 | ||||
| DQ148475.1 | ABA29340.1 |
More Information
Set Membership: |
|

Genomic Map