Assay Details
Target Gene Details
Entrez Gene ID: | 12716 |
Gene Name: | creatine kinase, mitochondrial 1, ubiquitous |
Gene Aliases: |
Mt-CK, ScCKmit, UbCKmit, mi-CK |
Location: |
Chr.2:121357751-121363737 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ckmt1 | NM_009897.2 | 9 | 1461 | NP_034027.1 |
| AK005076.1 | 9 | 1460 | BAB23800.1 | |
| AK131953.1 | 9 | 1465 | BAE20897.1 | |
| BC025976.1 | 10 | 1376 | AAH25976.1 | |
| Z13968.1 | 9 | 1461 | CAA78371.1 |
Target Gene Details
Entrez Gene ID: | 140476 |
Gene Name: | stereocilin |
Gene Aliases: |
DFNB16 |
Location: |
Chr.2:121363248-121381370 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Strc |
More Information
Set Membership: |
|

Genomic Map