Assay Details
Target Gene Details
Entrez Gene ID: | 58202 |
Gene Name: | negative elongation factor complex member B |
Gene Aliases: |
A730008L03Rik, AB041607, AI663983, Cobra1, Nelf-b |
Location: |
Chr.2:25199712-25211489 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nelfb | NM_021393.2 | 5 | 928 | NP_067368.2 |
| AB041607.1 | 4 | 1049 | BAA95090.1 | |
| AK080419.1 | 5 | 630 | ||
| AK087524.1 | 5 | 628 | ||
| AK173124.1 | 4 | 463 | BAD32402.1 | |
| AK177267.1 | 2 | 228 | ||
| AK184385.1 | 2 | 228 | ||
| AK198680.1 | ||||
| AK203337.1 | 3 | 310 | ||
| AK217288.1 | 2 | 266 | ||
| BC004762.1 | 5 | 816 | AAH04762.1 | |
| BU519080.1 | 5 | 861 |

Genomic Map