Assay Details
Target Gene Details
Entrez Gene ID: | 58202 |
Gene Name: | negative elongation factor complex member B |
Gene Aliases: |
A730008L03Rik, AB041607, AI663983, Cobra1, Nelf-b |
Location: |
Chr.2:25199712-25211489 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nelfb | NM_021393.2 | NP_067368.2 | ||
| AB041607.1 | BAA95090.1 | |||
| AK080419.1 | ||||
| AK087524.1 | ||||
| AK173124.1 | BAD32402.1 | |||
| AK179149.1 | ||||
| AK189417.1 | ||||
| AK214584.1 | ||||
| BC004762.1 | AAH04762.1 |

Genomic Map