Assay Details
Target Gene Details
Entrez Gene ID: | 66170 |
Gene Name: | coiled-coil-helix-coiled-coil-helix domain containing 5 |
Gene Aliases: |
1110027L01Rik, AW045710 |
Location: |
Chr.2:129129700-129134112 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chchd5 | NM_025395.3 | NP_079671.1 | ||
| AK003341.1 | BAB22728.1 | |||
| AK003936.1 | ||||
| AK013018.1 | BAB28600.1 | |||
| BC094414.1 | AAH94414.1 | |||
| BC096668.1 | AAH96668.1 |

Genomic Map