Assay Details
Target Gene Details
Entrez Gene ID: | 108699 |
Gene Name: | chimerin 1 |
Gene Aliases: |
0610007I19Rik, 0710001E19Rik, 1700112L09Rik, 2900046J01Rik, AI413815, ARHGAP2 |
Location: |
Chr.2:73610660-73775367 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chn1 | NM_001113246.2 | NP_001106717.2 | ||
| NM_001166603.1 | NP_001160075.1 | |||
| NM_001166604.1 | NP_001160076.1 | |||
| NM_175752.3 | NP_786928.2 | |||
| AB264771.1 | ||||
| AB264772.1 | ||||
| AF332069.1 | AAK56097.1 | |||
| AF332070.1 | AAK56098.1 | |||
| AK032630.1 | ||||
| AK049943.1 | BAC33996.1 | |||
| AK075606.1 | BAC35853.1 | |||
| AK147686.1 | BAE28074.1 | |||
| BC010825.1 | AAH10825.1 | |||
| BC024796.1 | AAH24796.1 | |||
| BC025023.1 | AAH25023.1 | |||
| BC054770.1 | AAH54770.1 |

Genomic Map