Assay Details
Target Gene Details
Entrez Gene ID: | 108699 |
Gene Name: | chimerin 1 |
Gene Aliases: |
0610007I19Rik, 0710001E19Rik, 1700112L09Rik, 2900046J01Rik, AI413815, ARHGAP2 |
Location: |
Chr.2:73610660-73775367 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chn1 | NM_001113246.2 | 12 | 1463 | NP_001106717.2 |
| NM_001166603.1 | 11 | 1289 | NP_001160075.1 | |
| NM_001166604.1 | 5 | 1101 | NP_001160076.1 | |
| NM_029716.3 | 5 | 664 | NP_083992.1 | |
| NM_175752.3 | 6 | 1275 | NP_786928.2 | |
| AB264771.1 | 5 | 925 | ||
| AB264772.1 | 11 | 986 | ||
| AB264774.1 | 4 | 1065 | ||
| AF332069.1 | 6 | 783 | AAK56097.1 | |
| AF332070.1 | 6 | 783 | AAK56098.1 | |
| AK007182.1 | 5 | 664 | BAB24888.1 | |
| AK032630.1 | 6 | 1233 | ||
| AK049943.1 | 6 | 1219 | BAC33996.1 | |
| AK075606.1 | 6 | 1237 | BAC35853.1 | |
| AK147686.1 | 6 | 1219 | BAE28074.1 | |
| BC010825.1 | 6 | 1250 | AAH10825.1 | |
| BC024796.1 | 6 | 1226 | AAH24796.1 | |
| BC025023.1 | 6 | 1206 | AAH25023.1 | |
| BC054770.1 | 12 | 1215 | AAH54770.1 |
Target Gene Details
Entrez Gene ID: | 329427 |
Gene Name: | chimerin 1, opposite strand 3 |
Gene Aliases: |
Gm13704, OTTMUSG00000013429 |
Location: |
Chr.2:73596526-73616012 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chn1os3 | NR_040623.1 | 4 | 1407 |

Genomic Map