Assay Details
Target Gene Details
Entrez Gene ID: | 56264 |
Gene Name: | carboxypeptidase X 1 (M14 family) |
Gene Aliases: |
AA986902, Cpx-1, Cpx1, Cpxm |
Location: |
Chr.2:130390775-130397629 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpxm1 | NM_019696.2 | 2 | 398 | NP_062670.2 |
| AF077738.1 | 2 | 343 | AAD15985.1 | |
| BC003713.1 | 2 | 331 | AAH03713.1 | |
| BY282671.1 | 2 | 398 |
More Information
Set Membership: |
|

Genomic Map