Assay Details
Target Gene Details
Entrez Gene ID: | 12995 |
Gene Name: | casein kinase 2, alpha 1 polypeptide |
Gene Aliases: |
Csnk2a1-rs4 |
Location: |
Chr.2:152226731-152281856 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csnk2a1 | NM_007788.3 | NP_031814.2 | ||
| AK146032.1 | BAE26845.1 | |||
| AK158077.1 | BAE34350.1 | |||
| BC026149.1 | AAH26149.1 | |||
| BC060742.1 | AAH60742.1 | |||
| BC089343.1 | AAH89343.1 | |||
| U17112.1 | AAA64563.1 | |||
| U51866.1 | AAA96795.1 |

Genomic Map