Assay Details
Target Gene Details
Entrez Gene ID: | 56431 |
Gene Name: | destrin |
Gene Aliases: |
2610043P17Rik, ADF, AU042046, Dsn, corn1, sid23p |
Location: |
Chr.2:143915331-143943324 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dstn | NM_019771.2 | NP_062745.1 | ||
| AB025406.1 | BAA84691.1 | |||
| AK078898.1 | BAC37447.1 | |||
| BC131926.1 | ||||
| CT010216.1 | CAJ18424.1 |
More Information
Set Membership: |
|

Genomic Map