Assay Details
Target Gene Details
Entrez Gene ID: | 52838 |
Gene Name: | DNL-type zinc finger |
Gene Aliases: |
1110034G11Rik, D2Bwg1335e, HEP1 |
Location: |
Chr.2:26348118-26352110 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnlz | NM_001139503.1 | NP_001132975.1 | ||
| NM_001139504.1 | 2 | 413 | NP_001132976.1 | |
| NM_026828.3 | 2 | 409 | NP_081104.1 | |
| AK004089.1 | 2 | 399 | BAB23162.1 | |
| AK142070.1 | 2 | 410 | BAE24931.1 | |
| BC031595.1 | AAH31595.1 | |||
| BC139333.1 | 2 | 373 | ||
| BC139334.1 | 2 | 372 | ||
| BY266852.1 |

Genomic Map