Assay Details
Target Gene Details
Entrez Gene ID: | 69185 |
Gene Name: | DTW domain containing 1 |
Gene Aliases: |
1810033A06Rik |
Location: |
Chr.2:126152113-126165277 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dtwd1 | NM_026981.2 | NP_081257.1 | ||
| AK149746.1 | ||||
| BC020149.1 | AAH20149.1 | |||
| BY240339.1 |

Genomic Map