Assay Details
Target Gene Details
Entrez Gene ID: | 23856 |
Gene Name: | death inducer-obliterator 1 |
Gene Aliases: |
6720461J16Rik, D130048F08Rik, DATF-1, DIO-1, Datf1, dido |
Location: |
Chr.2:180657963-180710035 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dido1 | NM_001291432.1 | NP_001278361.1 | ||
| NM_177852.4 | NP_808520.2 | |||
| AK051426.1 | 1 | 145 | ||
| AK129117.1 | BAC97927.1 | |||
| BC096662.1 | AAH96662.1 |
More Information
Set Membership: |
|

Genomic Map