Assay Details
Target Gene Details
Entrez Gene ID: | 140579 |
Gene Name: | engulfment and cell motility 2 |
Gene Aliases: |
1190002F24Rik, CED-12 |
Location: |
Chr.2:165288031-165326479 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elmo2 | NM_080287.2 | 18 | 1847 | NP_525026.2 |
| NM_207706.1 | 19 | 1883 | NP_997589.1 | |
| AB093301.1 | 9 | 2428 | BAC41483.3 | |
| AF398884.1 | 17 | 1746 | AAL14465.1 | |
| AK032033.1 | 19 | 1856 | BAC27662.1 | |
| AK033918.1 | 18 | 2041 | BAC28514.1 | |
| AK035710.1 | 3 | 233 | BAC29162.1 | |
| AK038097.1 | 18 | 1836 | ||
| AK038455.1 | 18 | 1953 | BAC30007.1 | |
| AK045428.1 | 18 | 1848 | BAC32361.1 | |
| AK047040.1 | 18 | 1809 | BAC32945.1 | |
| AK050823.1 | 18 | 1810 | BAC34424.1 | |
| AK053574.1 | 19 | 1847 | BAC35433.1 | |
| AK129023.1 | 18 | 1847 | ||
| AK205725.1 | 1 | 40 | ||
| BC023954.1 | 19 | 1868 | AAH23954.1 |
More Information
Set Membership: |
|

Genomic Map