Assay Details
Target Gene Details
Entrez Gene ID: | 140579 |
Gene Name: | engulfment and cell motility 2 |
Gene Aliases: |
1190002F24Rik, CED-12 |
Location: |
Chr.2:165288031-165326479 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elmo2 | NM_080287.2 | NP_525026.2 | ||
| NM_207706.1 | NP_997589.1 | |||
| AB093301.1 | BAC41483.3 | |||
| AF398884.1 | AAL14465.1 | |||
| AK004448.1 | BAB23307.1 | |||
| AK032033.1 | BAC27662.1 | |||
| AK033918.1 | BAC28514.1 | |||
| AK035710.1 | BAC29162.1 | |||
| AK038097.1 | ||||
| AK038455.1 | BAC30007.1 | |||
| AK045428.1 | BAC32361.1 | |||
| AK047040.1 | BAC32945.1 | |||
| AK050823.1 | BAC34424.1 | |||
| AK053574.1 | BAC35433.1 | |||
| AK129023.1 | ||||
| AK205725.1 | ||||
| BC023954.1 | AAH23954.1 |
More Information
Set Membership: |
|

Genomic Map