Assay Details
Target Gene Details
Entrez Gene ID: | 13537 |
Gene Name: | dual specificity phosphatase 2 |
Gene Aliases: |
PAC1 |
Location: |
Chr.2:127336159-127338377 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dusp2 | NM_010090.2 | NP_034220.2 | ||
| AK088059.1 | ||||
| AK134067.1 | BAE21999.1 | |||
| BC048696.1 | AAH48696.1 | |||
| L11330.1 | AAA19666.1 |
More Information
Set Membership: |
|

Genomic Map