Assay Details
Target Gene Details
Entrez Gene ID: | 228598 |
Gene Name: | early B cell factor 4 |
Gene Aliases: |
Ebf3, O/E-4, Olf-1 |
Location: |
Chr.2:130295249-130370481 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ebf4 | NM_001110513.1 | NP_001103983.1 | ||
| AF387630.1 | AAM97580.1 | |||
| AF387631.1 | AAM97581.1 | |||
| AF387632.1 | AAM97582.1 | |||
| AF387633.1 | AAM97583.1 | |||
| AF387634.1 | AAM97584.1 | |||
| BC138119.1 | ||||
| BC145155.1 |
More Information
Set Membership: |
|

Genomic Map