Assay Details
Target Gene Details
Entrez Gene ID: | 320132 |
Gene Name: | RIKEN cDNA A230108P19 gene |
Gene Aliases: |
- |
Location: |
Chr.2:6193251-6321611 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A230108P19Rik | NR_040333.1 | |||
| AK039218.1 | ||||
| AK044786.1 |
Target Gene Details
Entrez Gene ID: | 67856 |
Gene Name: | enoyl Coenzyme A hydratase domain containing 3 |
Gene Aliases: |
2310005D12Rik, AI662097 |
Location: |
Chr.2:6188465-6212994 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Echdc3 | NM_024208.4 | 4 | 524 | NP_077170.2 |
| AK004218.1 | 4 | 526 | ||
| AK009166.1 | 4 | 525 | BAB26117.1 | |
| AK014736.1 | 4 | 552 | ||
| BC002214.1 | 4 | 505 | AAH02214.1 | |
| BC054365.1 | 4 | 493 | AAH54365.1 |
More Information
Set Membership: |
|

Genomic Map