Assay Details
Target Gene Details
Entrez Gene ID: | 269344 |
Gene Name: | elongation factor RNA polymerase II-like 3 |
Gene Aliases: |
A930015D22Rik |
Location: |
Chr.2:121439025-121444278 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ell3 | NM_145973.2 | NP_666085.2 | ||
| AK139676.1 | ||||
| BC045151.1 | AAH45151.1 |
More Information
Set Membership: |
|

Genomic Map