Assay Details
Target Gene Details
Entrez Gene ID: | 67809 |
Gene Name: | regulator of microtubule dynamics 3 |
Gene Aliases: |
1200015F23Rik, AI131757, Fam82a2, Ptpip51, RMD-3, Rmd3 |
Location: |
Chr.2:119136998-119157034 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rmdn3 | NM_001033136.3 | NP_001028308.1 | ||
| AK004786.1 | ||||
| AK146299.1 | BAE27056.1 | |||
| AK151090.1 | BAE30103.1 | |||
| AK153322.1 | BAE31902.1 | |||
| AK153369.1 | BAE31937.1 | |||
| AK171770.1 | ||||
| BC055754.1 | AAH55754.1 |

Genomic Map