Assay Details
Target Gene Details
Entrez Gene ID: | 71405 |
Gene Name: | family with sequence similarity 83, member C |
Gene Aliases: |
5530400B04Rik |
Location: |
Chr.2:155829183-155834854 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam83c | NM_027788.2 | NP_082064.2 | ||
| BC113190.1 | ||||
| BC113797.1 |

Genomic Map