Assay Details
Target Gene Details
Entrez Gene ID: | 208618 |
Gene Name: | enhancer trap locus 4 |
Gene Aliases: |
6620402G01Rik, 9430077C05Rik, BC026657, E330027G05Rik, Etl-4, Skt |
Location: |
Chr.2:19909763-20810535 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Etl4 | NM_001081006.1 | 2 | 262 | NP_001074475.1 |
| AK052020.1 | BAC34833.1 | |||
| CF744489.1 | 2 | 262 |

Genomic Map