Assay Details
Target Gene Details
Entrez Gene ID: | 208618 |
Gene Name: | enhancer trap locus 4 |
Gene Aliases: |
6620402G01Rik, 9430077C05Rik, BC026657, E330027G05Rik, Etl-4, Skt |
Location: |
Chr.2:19909763-20810535 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Etl4 | NM_001081006.1 | 5 | 899 | NP_001074475.1 |
| NM_001177630.2 | 4 | 997 | NP_001171101.2 | |
| NM_001177631.1 | 4 | 997 | NP_001171102.1 | |
| NM_029895.4 | 5 | 1210 | NP_084171.2 | |
| NM_178059.5 | 5 | 1210 | NP_835160.2 | |
| AB093289.2 | 4 | 787 | BAC41473.2 | |
| AB125594.2 | 5 | 1210 | BAD14929.1 | |
| AB125595.2 | 5 | 1210 | BAD14930.1 | |
| AK052020.1 | 6 | 1312 | BAC34833.1 | |
| AK052733.1 | 4 | 685 | BAC35121.1 | |
| AK147510.1 | 4 | 904 | ||
| BC050016.1 | 4 | 700 | AAH50016.1 | |
| BC158051.1 | 4 | 719 |

Genomic Map