Assay Details
Target Gene Details
Entrez Gene ID: | 12496 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 2 |
Gene Aliases: |
Cd39l1, NTPDase2 |
Location: |
Chr.2:25395874-25401323 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Entpd2 | NM_009849.2 | NP_033979.2 | ||
| AF042811.1 | AAC24347.1 | |||
| AK002553.1 | BAB22182.1 | |||
| AK172281.1 | BAE42922.1 | |||
| AY376711.1 | AAQ86586.1 | |||
| BC011241.1 | AAH11241.1 | |||
| BY237099.1 | ||||
| U91511.1 | AAB81014.1 |
More Information
Set Membership: |
|

Genomic Map