Assay Details
Target Gene Details
Entrez Gene ID: | 68126 |
Gene Name: | fumarylacetoacetate hydrolase domain containing 2A |
Gene Aliases: |
1500003K14Rik, B430104H02Rik, Cgi-105, Fahd2 |
Location: |
Chr.2:127436215-127444565 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fahd2a | NM_029629.2 | NP_083905.1 | ||
| AI841210.1 | ||||
| AK002291.1 | ||||
| AK005134.1 | ||||
| AK080868.1 | BAC38052.1 | |||
| AK170875.1 | BAE42085.1 | |||
| BC030182.1 | AAH30182.1 |
More Information
Set Membership: |
|

Genomic Map