Assay Details
Target Gene Details
Entrez Gene ID: | 98952 |
Gene Name: | family with sequence similarity 102, member A |
Gene Aliases: |
AI426465, C230093N12Rik, Eeig1 |
Location: |
Chr.2:32535359-32569750 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam102a | NM_153560.4 | NP_705788.1 | ||
| AK049032.1 | BAC33519.1 | |||
| AK131145.1 | BAD21395.1 | |||
| AK157331.1 | BAE34050.1 | |||
| AK157490.1 | BAE34101.1 | |||
| AK158435.1 | ||||
| AK165751.1 | ||||
| AK170536.1 | BAE41864.1 | |||
| AK170847.1 | BAE42070.1 | |||
| AK170951.1 | BAE42135.1 | |||
| AK172536.1 | BAE43056.1 | |||
| AK186631.1 | ||||
| BC023752.1 | AAH23752.1 | |||
| BC031157.1 | AAH31157.1 |

Genomic Map