Assay Details
Target Gene Details
Entrez Gene ID: | 72543 |
Gene Name: | multivesicular body subunit 12B |
Gene Aliases: |
2610200O14Rik, 2610528K11Rik, AI414895, Fam125b |
Location: |
Chr.2:33729953-33887946 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mvb12b | NM_175184.4 | NP_780393.2 | ||
| AK012178.1 | BAC25358.1 | |||
| AK087821.1 | BAC40015.1 | |||
| AK131113.1 | BAD21363.1 | |||
| AK135990.1 | ||||
| AK154267.1 | ||||
| BC049129.1 | AAH49129.2 | |||
| BC059907.1 | AAH59907.1 |

Genomic Map