Assay Details
Target Gene Details
Entrez Gene ID: | 227737 |
Gene Name: | family with sequence similarity 129, member B |
Gene Aliases: |
9130404D14Rik |
Location: |
Chr.2:32876134-32925253 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam129b | NM_146119.2 | NP_666231.1 | ||
| AK033735.1 | BAC28456.1 | |||
| AK046583.1 | BAC32795.1 | |||
| AK051396.1 | BAC34625.1 | |||
| AK146953.1 | BAE27562.1 | |||
| AK149818.1 | ||||
| AK149888.1 | BAE29147.1 | |||
| AK150642.1 | BAE29730.1 | |||
| AK152207.1 | BAE31035.1 | |||
| AK153489.1 | BAE32037.1 | |||
| AK170512.1 | BAE41849.1 | |||
| BC027843.1 | AAH27843.1 |

Genomic Map