Assay Details
Target Gene Details
Entrez Gene ID: | 68222 |
Gene Name: | family with sequence similarity 166, member A |
Gene Aliases: |
4931415M17 |
Location: |
Chr.2:25214724-25222281 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam166a | NM_026624.3 | 7 | 978 | NP_080900.1 |
| AK016459.1 | 7 | 980 | BAB30248.1 | |
| BC061039.1 | 7 | 981 | AAH61039.1 | |
| BX631039.1 | 1 | 153 |
Target Gene Details
Entrez Gene ID: | 227613 |
Gene Name: | tubulin, beta 4B class IVB |
Gene Aliases: |
4930542G03Rik, Tubb2c, Tubb2c1 |
Location: |
Chr.2:25222158-25224702 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubb4b | NM_146116.2 | NP_666228.1 | ||
| AK002568.1 | BAB22193.2 | |||
| AK015901.1 | ||||
| AK076895.1 | ||||
| AK146587.1 | BAE27282.1 | |||
| AK167022.1 | BAE39195.1 | |||
| AK194925.1 | ||||
| AK214079.1 | ||||
| BC005547.1 | AAH05547.1 | |||
| BC022919.1 | AAH22919.1 | |||
| BC083319.1 | AAH83319.1 | |||
| CJ309661.1 |
More Information
Set Membership: |
|

Genomic Map