Assay Details
Target Gene Details
Entrez Gene ID: | 241520 |
Gene Name: | family with sequence similarity 171, member B |
Gene Aliases: |
D430039N05Rik |
Location: |
Chr.2:83812728-83883486 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam171b | NM_175514.2 | NP_780723.1 | ||
| AK038899.1 | BAC30163.1 | |||
| AK052508.1 | BAC35021.1 | |||
| BC113787.1 |

Genomic Map