Assay Details
Target Gene Details
Entrez Gene ID: | 98970 |
Gene Name: | fibrinogen C domain containing 1 |
Gene Aliases: |
AI448887 |
Location: |
Chr.2:31813290-31846647 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fibcd1 | NM_178887.4 | 4 | 1005 | NP_849218.2 |
| AK046338.1 | 4 | 967 | BAC32685.1 | |
| AK084379.1 | 5 | 1008 | BAC39169.1 | |
| BC060634.1 | 4 | 1005 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map