Assay Details
Target Gene Details
Entrez Gene ID: | 320267 |
Gene Name: | far upstream element (FUSE) binding protein 3 |
Gene Aliases: |
A330051M14Rik, AV006371, FBP3, Marta2 |
Location: |
Chr.2:31572651-31617526 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fubp3 | NM_001033389.4 | NP_001028561.2 | ||
| NM_001290548.1 | NP_001277477.1 | |||
| AK145377.1 | BAE26399.1 | |||
| AK166081.1 | ||||
| AK166478.1 | ||||
| AK167671.1 | BAE39720.1 | |||
| BG087512.2 |
More Information
Set Membership: |
|

Genomic Map