Assay Details
Target Gene Details
Entrez Gene ID: | 14133 |
Gene Name: | ficolin A |
Gene Aliases: |
Fcn1 |
Location: |
Chr.2:25624666-25635443 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fcna | NM_007995.3 | NP_032021.1 | ||
| AB007813.1 | BAA25126.1 | |||
| AB222271.1 | BAE53381.1 | |||
| AK133539.1 | BAE21714.1 | |||
| AK138752.1 | BAE23769.1 | |||
| AK159651.1 | BAE35262.1 | |||
| BC019180.1 | AAH19180.1 | |||
| CT010383.1 | CAJ18590.1 |
More Information
Set Membership: |
|

Genomic Map