Assay Details
Target Gene Details
Entrez Gene ID: | 99326 |
Gene Name: | GTPase activating RANGAP domain-like 3 |
Gene Aliases: |
AW120551 |
Location: |
Chr.2:32986366-33131888 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Garnl3 | NM_178888.4 | NP_849219.2 | ||
| AK034068.1 | BAC28570.1 | |||
| AK036258.1 | ||||
| AK083673.1 | BAC38990.1 | |||
| AK133163.1 | BAE21537.1 | |||
| BC132143.1 |
More Information
Set Membership: |
|

Genomic Map