Assay Details
Target Gene Details
Entrez Gene ID: | 320415 |
Gene Name: | GTP cyclohydrolase I feedback regulator |
Gene Aliases: |
2010323F13Rik, GFRP, P35 |
Location: |
Chr.2:119167788-119172389 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gchfr | NM_177157.4 | NP_796131.1 | ||
| AK028116.1 | BAC25756.1 | |||
| AK028122.1 | BAC25761.1 | |||
| BC096406.1 | AAH96406.1 | |||
| BC132450.1 | ||||
| BC132478.1 |
More Information
Set Membership: |
|

Genomic Map