Assay Details
Target Gene Details
Entrez Gene ID: | 69270 |
Gene Name: | GINS complex subunit 1 (Psf1 homolog) |
Gene Aliases: |
2810418N01Rik, Gins4, PSF1, mKIAA0186 |
Location: |
Chr.2:150909557-150931280 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gins1 | NM_001163476.1 | 4 | 408 | NP_001156948.1 |
| NM_027014.1 | 4 | 408 | NP_081290.1 | |
| AK013116.1 | 4 | 401 | BAB28658.1 | |
| AK129081.1 | 3 | 199 | BAC97891.1 | |
| BC027537.1 | 4 | 385 | AAH27537.1 | |
| BQ553304.1 | 4 | 408 |
More Information
Set Membership: |
|

Genomic Map